Lab coat, Non-Sterile No Pocket polypropylene fabrics, Light Weight, easy breathability, knee length, Velcro front, elastic cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, Orange, 2XL
Catalog #: 76488-476
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Customers Who Bought This Also Bought