Lab coat, Easy-Breathe, Non-sterile, No pockets, Level 2, SMS fabrics, fluid resistant, easy breathability, knee length, snap front, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, White, S
Catalog #: 76488-450
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Customers Who Bought This Also Bought