Lab coat, Extra-safe, Non-sterile, No pockets, Level 3, SMS fabrics, highly fluid resistant, knee length, snap front, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, Medical Blue, M
Catalog #: 76488-440
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Customers Who Bought This Also Bought