Lab coat, LiquidGuard, Non-sterile, No pockets, microporous fabrics, pass blood penetration test, pass ChemoDrug Permeation Test, knee length, snap front, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, White, S
Catalog #: 76488-412
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Customers Who Bought This Also Bought