Coverall, Sterile, LiquidGuard, attached hood, attached booties, pass blood penetration test, breathable, low particle shedding, rated category 1 per Helmke Drum Test, manufactured in controlled environment, Gamma Irradiated validated sterile to SAL 10-6, White, 3XL
Catalog #: 76488-304
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Customers Who Bought This Also Bought