Filter Cover, NxMD White, For N95 Filter Pads. Suitable For COVID virus in accordance with CDC guidelines and Non oil based aerosols. Can be mounted directly on all series half and full face masks
Catalog #: CA76488-150
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought