Filter Cover and holder, NxMD White, with N95 Filters. Suitable For COVID virus in accordance with CDC guidelines and Non oil based aerosols. Can be mounted directly on all series half and full face masks. (20 Pair/case)
Catalog #: CA76488-142
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought