Respirator, NxMD Half Mask, with Filter Cover and Holder. No Exhalation valve. Elastomeric. White. M/L. NIOSH. Osha silica dust compliant when used with a N95 or P100 filter. Protection - N95 Non oil base particulate aerosols and COVID Viruses.
Catalog #: CA76488-130
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought