Respirator, Nx Half Mask, with N95 Filter, Cover & Holder. No Exhalation valve. Elastomeric. Black. M/L. NIOSH. Osha silica dust compliant when used with a N95 or P100 filter. Protection - N95 Non oil base particulate aerosols and COVID Viruses.
Catalog #: CA76488-118
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought