Air Monitor Badge, sampling involves no liquid and no mechanical pumps. The vapor sample difuses through a porous membrane and is collected on a specially prepared 2,4-DNPH coated high-purity silica gel adsorbent, clip the badge near the breathing zone to measure personal exposure
Catalog #: 76490-530
Supplier: CANCER DIAGNOSTICS, INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.