Biopsy Pad (Foam Black) Special thin non-stick foam (1in x 1.125in) allows for optimal fluid transfer with virtually no carryover. Pressure on biopsy is greatly reduced decreasing occurrence of artifact
Catalog #: 76490-514
Supplier: CANCER DIAGNOSTICS, INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.