Mask, Double-Seal Arching, White, pinch point bar on nose, chin and middle bar for comfort, breathability, better protection. Hypoallergenic for sensitive skin, softest ear loops, ultra-high filtration, PFE 99.7%, at 0.1micron, BFE 99.5%, very breathable. Latex free. ASTM Level 2 standard, 120mmHg
Catalog #: 76490-490
Supplier: CANCER DIAGNOSTICS, INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.