Order Entry
ContactUsLinkComponent
Bottle, Amber Boston Round with 24-400 Black Phenolic PolyCone Lined Cap attached, Vacuum & Ionized, 8oz (240ml
Boston Round Bottles, Vacuum and Ionized, Amber, Narrow Mouth, Qorpak
Catalog #: 76487-276
Supplier:  Qorpak
Bottle, Amber Boston Round with 24-400 Black Phenolic PolyCone Lined Cap attached, Vacuum & Ionized, 8oz (240ml
Boston Round Bottles, Vacuum and Ionized, Amber, Narrow Mouth, Qorpak
Catalog #: 76487-276
Supplier:  Qorpak

Specifications

  • Cap Size:
    24-400
  • Cap Type:
    Polycone lined black phenolic cap
  • Capacity:
    237 ml (8 oz.)
  • Cat. no.:
    76487-276

Specifications

About this item

The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.

  • Boston round bottles are made of glass

Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.

Customers Who Bought This Also Bought