Order Entry
ContactUsLinkComponent
Vial, Amber Borosilicate with 13-425 Green Thermoset F217 & PTFE Lined Cap attached, Cleaned for Volatiles15 x 45mm 1 dram (4ml
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Catalog #: 76487-246
Supplier:  Qorpak
Vial, Amber Borosilicate with 13-425 Green Thermoset F217 & PTFE Lined Cap attached, Cleaned for Volatiles15 x 45mm 1 dram (4ml
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Catalog #: 76487-246
Supplier:  Qorpak

Specifications

  • Cap included:
    Yes
  • Cap Size:
    13-425
  • Capacity:
    4.00 ml
  • Color:
    Amber
  • Description:
    Cleaned for volatiles
  • Cat. no.:
    76487-246
  • Diameter ×H:
    15×45 mm

Specifications

About this item

Amber screw thread sample vials offer protection from harmful light and UV rays for light sensitive products.

These general purpose glass vials are made of Type I borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.

Ordering information: Vials 36319-407 and 66175-540 are supplied pre-cleaned for volatiles. Vial 11311-522 is supplied ultra cleaned. Vials 93000-912, 93000-922, 93000-906, and 93000-928 are supplied vacuumed and ionized.
Customers Who Bought This Also Bought