Order Entry
ContactUsLinkComponent
Packer Bottle, Amber Wide Mouth with 28-400 Black Polypropylene Cap & PTFE Disc attached, Cleaned & Certified for Semi-Volatiles1oz (30ml
Packer Bottles, Amber, Wide Mouth
Catalog #: 76487-192
Supplier:  Qorpak
Packer Bottles, Amber, Wide Mouth
Catalog #: 76487-192
Supplier:  Qorpak

Specifications

  • Cap Size:
    28-400
  • Capacity:
    30 ml (1 oz.)
  • Cat. no.:
    76487-192

Specifications

About this item

Amber wide mouth Packer bottles feature a wide opening making them the ideal general-purpose bottle for solids.

  • Packer bottles are made of glass
  • Ideal for light sensitive products

Applications include pharmaceutical, chemical, and environmental.

Ordering information: Bottles 11311-528, 11311-530, 11311-532, 11311-534, 11311-540, 10158-494 and 10158-496 are supplied Ultra Cleaned. Bottles 36319-156, 36319-158, 36319-250, 36319-253, 36319-256, 36319-265, 36319-268, and are supplied pre-cleaned for volatiles. Bottles 89204-848, BA11311-362CRC, 76487-192, 76487-194, 76487-196, 76487-198 and 76487-200 are supplied pre-cleaned and certified for semi-volatiles.
Customers Who Bought This Also Bought