SFB Expansion, Oxford Nanopore Technologies
Catalog #: 76487-136
Supplier: Oxford Nanopore Technologies
CAS Number:
Specifications
- Compatible with:R9.4.1 flow cells
- Description:SFB expansion
- Cat. no.:76487-136
- No. of Reactions:4
Specifications
About this item
Short Fragment Buffer (SFB).
This kit is recommended for users who require additional Short Fragment Buffer for use with the 'PCR tiling of COVID-19 virus' protocol or any other experiment sequencing short fragments.
SFB Expansion contains four vials of Short Fragment Buffer (SFB), sufficient for four reactions.
Delivery: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Customers Who Bought This Also Bought