ContactUsLinkComponent
Buoffant Cap, White, Disposable spunbond polypropylene, Lightweight fabric, Elastic band for flexible fit. Latex free, protection against non-hazardous dry particulates like dirt, One time use, Size: 24in
Catalog #: 76486-988
Supplier:  PIP
Buoffant Cap, White, Disposable spunbond polypropylene, Lightweight fabric, Elastic band for flexible fit. Latex free, protection against non-hazardous dry particulates like dirt, One time use, Size: 24in
Catalog #: 76486-988
Supplier:  PIP
Customers Who Bought This Also Bought