ContactUsLinkComponent
Apron, Single use, Excellent resistance to chemicals, fats, oils and grease, Easy to clean, Convenient individual packaging, Die cut for consistent sizing, for light to medium duty applications, 1.5mil polyethylene, White, non-embossed, 28x55in
Catalog #: 76486-942
Supplier:  PIP
Apron, Single use, Excellent resistance to chemicals, fats, oils and grease, Easy to clean, Convenient individual packaging, Die cut for consistent sizing, for light to medium duty applications, 1.5mil polyethylene, White, non-embossed, 28x55in
Catalog #: 76486-942
Supplier:  PIP
Customers Who Bought This Also Bought