ContactUsLinkComponent
Coverall with Hood & Boot, M3, Grey, Five layers of protection providing a high level of breathability and protection against dry particulates, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex free, One time use, Size: XL
Catalog #: 76486-930
Supplier:  PIP
Coverall with Hood & Boot, M3, Grey, Five layers of protection providing a high level of breathability and protection against dry particulates, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex free, One time use, Size: XL
Catalog #: 76486-930
Supplier:  PIP
Customers Who Bought This Also Bought