ContactUsLinkComponent
Coverall with Hood, Elastic Wrists & Ankles, M3, Five layers of protection providing a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex free, One time use, Size: XL
Catalog #: 76486-918
Supplier:  PIP
Coverall with Hood, Elastic Wrists & Ankles, M3, Five layers of protection providing a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex free, One time use, Size: XL
Catalog #: 76486-918
Supplier:  PIP
Customers Who Bought This Also Bought