ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle, M3, Gray, Five layers of protection providing a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex Free, One time use, Size: 6XL
Catalog #: 76486-898
Supplier:  PIP
Coverall with Elastic Wrist & Ankle, M3, Gray, Five layers of protection providing a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex Free, One time use, Size: 6XL
Catalog #: 76486-898
Supplier:  PIP
Customers Who Bought This Also Bought