ContactUsLinkComponent
Coverall with Hood Elastic Wrist & Ankle, SMS, White, Full length zipper front, exceptional strength for easy donning, Serged seams Latex Free, One time use, 3 layers of protection for high breathability, Protection against dry particulates, water-based light liquid sprays and splashes, Size: 5XL
Catalog #: 76486-864
Supplier:  PIP
Coverall with Hood Elastic Wrist & Ankle, SMS, White, Full length zipper front, exceptional strength for easy donning, Serged seams Latex Free, One time use, 3 layers of protection for high breathability, Protection against dry particulates, water-based light liquid sprays and splashes, Size: 5XL
Catalog #: 76486-864
Supplier:  PIP
Customers Who Bought This Also Bought