ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle, White 45g SMS Coverall, zipper front, ANSI Sized for a wide range of motion, Serged seams, Latex Free, SMS technology provides three layers of protection for a high level of breathability, ANSI Sized for a wide range of motion, One time use, Size: 2xl
Catalog #: 76486-844
Supplier:  PIP
Coverall with Elastic Wrist & Ankle, White 45g SMS Coverall, zipper front, ANSI Sized for a wide range of motion, Serged seams, Latex Free, SMS technology provides three layers of protection for a high level of breathability, ANSI Sized for a wide range of motion, One time use, Size: 2xl
Catalog #: 76486-844
Supplier:  PIP
Customers Who Bought This Also Bought