ContactUsLinkComponent
Coverall, SMS - Basic, White, 45g, SMS, zipper front and collar, three layers of protection for a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Serged seams, Latex free, One time use, ANSI Sized for a wide range of motion, Size: 2xl
Catalog #: 76486-832
Supplier:  PIP
Coverall, SMS - Basic, White, 45g, SMS, zipper front and collar, three layers of protection for a high level of breathability and protection, Full length zipper front and exceptional strength for easy donning, Serged seams, Latex free, One time use, ANSI Sized for a wide range of motion, Size: 2xl
Catalog #: 76486-832
Supplier:  PIP
Customers Who Bought This Also Bought