ContactUsLinkComponent
Lab Coat - No Pockets, M3, White, SMMMS Snap Front Closure, 5 layers of protection providing high level of breathability, protection, against dry particulates, Full Length coverage to top of thigh, Snap button for quick donning, V neck design, Side slits for pocket access One time use
Catalog #: 76486-786
Supplier:  PIP
Lab Coat - No Pockets, M3, White, SMMMS Snap Front Closure, 5 layers of protection providing high level of breathability, protection, against dry particulates, Full Length coverage to top of thigh, Snap button for quick donning, V neck design, Side slits for pocket access One time use
Catalog #: 76486-786
Supplier:  PIP
Customers Who Bought This Also Bought