ContactUsLinkComponent
Shirt, M3, 50GSM, 100% FR Cotton Sateen, Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes, Snap buttons for quick donning, Professional collar, Serged seams, One time use, Size: M
Catalog #: 76486-770
Supplier:  PIP
Shirt, M3, 50GSM, 100% FR Cotton Sateen, Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes, Snap buttons for quick donning, Professional collar, Serged seams, One time use, Size: M
Catalog #: 76486-770
Supplier:  PIP
Customers Who Bought This Also Bought