ContactUsLinkComponent
Pants with Elastic Waist, M3, Latex Free Elastic waistband for comfort, Wide leg opening for easy donning, Designed for a roomy fit, Serged seams, 1 time use, 5 layers of protection providing high level of breathability, protection against dry particulate, water based light liquid spray, Size: m
Catalog #: 76486-756
Supplier:  PIP
Pants with Elastic Waist, M3, Latex Free Elastic waistband for comfort, Wide leg opening for easy donning, Designed for a roomy fit, Serged seams, 1 time use, 5 layers of protection providing high level of breathability, protection against dry particulate, water based light liquid spray, Size: m
Catalog #: 76486-756
Supplier:  PIP
Customers Who Bought This Also Bought