ContactUsLinkComponent
Sleeves, PosiWear M3, Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes, Elastic openings designed to fit most sizes, White in color, 18in length to fit most arm lengths
Catalog #: 76486-750
Supplier:  PIP
Sleeves, PosiWear M3, Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes, Elastic openings designed to fit most sizes, White in color, 18in length to fit most arm lengths
Catalog #: 76486-750
Supplier:  PIP
Customers Who Bought This Also Bought