ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle, M3, Elastic Wrist and Ankles keeps dirt and debris out, Full length zipper front and exceptional strength for easy donning, White SMMMS, Serged seams, Elastic waistband for improved fit, Latex Free, high level of breathability and protection, Size: 3xl
Catalog #: 76486-716
Supplier:  PIP
Coverall with Elastic Wrist & Ankle, M3, Elastic Wrist and Ankles keeps dirt and debris out, Full length zipper front and exceptional strength for easy donning, White SMMMS, Serged seams, Elastic waistband for improved fit, Latex Free, high level of breathability and protection, Size: 3xl
Catalog #: 76486-716
Supplier:  PIP
Customers Who Bought This Also Bought