Coverall, M3, Basic, White SMMMS, collar, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex Free, One time use, Five layers of protection providing a high level of breathability, protection against dry particulates, Size: 4xl
Catalog #: 76486-700
Supplier: PIP
Customers Who Bought This Also Bought