ContactUsLinkComponent
Coverall, M3, Basic, White SMMMS, collar, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex Free, One time use, Five layers of protection providing a high level of breathability, protection against dry particulates, Size: L
Catalog #: 76486-690
Supplier:  PIP
Coverall, M3, Basic, White SMMMS, collar, Full length zipper front and exceptional strength for easy donning, Elastic waistband for improved fit, Serged seams, Latex Free, One time use, Five layers of protection providing a high level of breathability, protection against dry particulates, Size: L
Catalog #: 76486-690
Supplier:  PIP
Customers Who Bought This Also Bought