ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle, SMS, Blue, 45g, Three layers of protection for high breathability, Protection against dry particulates, Protection against water-based ligh liquid sprays/splashes, Zipper front, Serged Seams, Latex Free, One time use, Size: 5xl
Catalog #: 76486-666
Supplier:  PIP
Coverall with Elastic Wrist & Ankle, SMS, Blue, 45g, Three layers of protection for high breathability, Protection against dry particulates, Protection against water-based ligh liquid sprays/splashes, Zipper front, Serged Seams, Latex Free, One time use, Size: 5xl
Catalog #: 76486-666
Supplier:  PIP
Customers Who Bought This Also Bought