ContactUsLinkComponent
Boot Cover, White, Ultimate Barrier, against dry particulates and liquid splash including viral and blood borne pathogens, Roomy construction to fit 18in boots, Elastic band at top of boots, Latex Free, Base material: Polyethylene/Polypropylene, One time use, Size: one size
Catalog #: 76486-626
Supplier:  PIP
Boot Cover, White, Ultimate Barrier, against dry particulates and liquid splash including viral and blood borne pathogens, Roomy construction to fit 18in boots, Elastic band at top of boots, Latex Free, Base material: Polyethylene/Polypropylene, One time use, Size: one size
Catalog #: 76486-626
Supplier:  PIP
Customers Who Bought This Also Bought