Coverall with Elastic Wrist & Ankle, Attached Hood & Boot, UB, White, Full length zipper front for easy donning, Elastic waistband for improved fit, Latex Free, PolySafe material which provide highest level of barrier protection against chemical, dry particulate, light spray, liquid splash, Size: xl
Catalog #: 76486-612
Supplier: PIP
Customers Who Bought This Also Bought