ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle, Attached Hood & Boot, UB Plus, Yellow, zipper front w/elastic wrist/ankle, attached hood, Elastic waistband for improved fit, Bound seams for improved protection, Storm flap with adhesive strip helps protection against intrusion, Latex Free, Size: 3xl
Catalog #: 76486-552
Supplier:  PIP
Coverall with Elastic Wrist & Ankle, Attached Hood & Boot, UB Plus, Yellow, zipper front w/elastic wrist/ankle, attached hood, Elastic waistband for improved fit, Bound seams for improved protection, Storm flap with adhesive strip helps protection against intrusion, Latex Free, Size: 3xl
Catalog #: 76486-552
Supplier:  PIP
Customers Who Bought This Also Bought