ContactUsLinkComponent
Coverall Hood & Boot, Microporous, zipper front with Elastic Wrist/Ankle, Polypropylene/Film Laminate, White, Breathable microporous material that protects against dry particulates and liquid sprays, Full length zipper front for easy donning, Low linting, Latex Free, One time use, Size: xl
Catalog #: 76486-534
Supplier:  PIP
Coverall Hood & Boot, Microporous, zipper front with Elastic Wrist/Ankle, Polypropylene/Film Laminate, White, Breathable microporous material that protects against dry particulates and liquid sprays, Full length zipper front for easy donning, Low linting, Latex Free, One time use, Size: xl
Catalog #: 76486-534
Supplier:  PIP
Customers Who Bought This Also Bought