ContactUsLinkComponent
Lab Coat - 2 Pocket, BA (Breathable Advantage) against dry particulates, Snap front with chest and hip pockets for extra storage, Full length coverage to thigh, Extremely low linting, Latex Free, ASTM 1670 standard for blood penetration, NFPA 99 Antistatic test, One time use, Size:
Catalog #: 76486-470
Supplier:  PIP
Lab Coat - 2 Pocket, BA (Breathable Advantage) against dry particulates, Snap front with chest and hip pockets for extra storage, Full length coverage to thigh, Extremely low linting, Latex Free, ASTM 1670 standard for blood penetration, NFPA 99 Antistatic test, One time use, Size:
Catalog #: 76486-470
Supplier:  PIP
Customers Who Bought This Also Bought