ContactUsLinkComponent
Shirt, Microporous White, BA (Breathable Advantage) against dry particulates and liquid sprays, Snap buttons for quick donning, Professional collar, ASTM 1670 standard for blood penetration, NFPA 99 Antistatic test, Extremely low linting, One time use, Size: 3XL
Catalog #: 76486-460
Supplier:  PIP
Shirt, Microporous White, BA (Breathable Advantage) against dry particulates and liquid sprays, Snap buttons for quick donning, Professional collar, ASTM 1670 standard for blood penetration, NFPA 99 Antistatic test, Extremely low linting, One time use, Size: 3XL
Catalog #: 76486-460
Supplier:  PIP
Customers Who Bought This Also Bought