Boot Cover, BA, (Breathable Advantage), Microporous, made from PolyBreathe light material, Elastic band at the top of shoe, NFPA 99 Antistatic test, ASTM 1670 standard for blood penetration, Extremely low linting, Latex free, One time use, Size: one size
Catalog #: 76486-444
Supplier: PIP
Customers Who Bought This Also Bought