ContactUsLinkComponent
Coverall with Hood, Elastic Wrist & Ankle, BA, against dry particulates, liquid sprays without sacrificing breathability, Full length zipper front and strength for easy donning, ANSI Sized for wide range of motion, Elastic waistband for improved fit, Extremely low linting, Latex free, Size: 2xl
Catalog #: 76486-410
Supplier:  PIP
Coverall with Hood, Elastic Wrist & Ankle, BA, against dry particulates, liquid sprays without sacrificing breathability, Full length zipper front and strength for easy donning, ANSI Sized for wide range of motion, Elastic waistband for improved fit, Extremely low linting, Latex free, Size: 2xl
Catalog #: 76486-410
Supplier:  PIP
Customers Who Bought This Also Bought