ContactUsLinkComponent
Shoe Cover, White, SBP, Standard Weight, Spunbond Polypropylene offers lightweight protection against dry particulates, Roomy construction to fit most shoe sizes, Elastic band at top of shoe to keep dirt off or on the shoe, One time use, Size: One size
Catalog #: 76486-350
Supplier:  PIP
Shoe Cover, White, SBP, Standard Weight, Spunbond Polypropylene offers lightweight protection against dry particulates, Roomy construction to fit most shoe sizes, Elastic band at top of shoe to keep dirt off or on the shoe, One time use, Size: One size
Catalog #: 76486-350
Supplier:  PIP
Customers Who Bought This Also Bought