ContactUsLinkComponent
Lab Coat with No Pocket, SBP, Blue, polypropylene disposable clothing provides affordable, lightweight protection against non-hazardous dry particulate like dirt, Professional collar, Full Length coverage to top of thigh, Snap button for front for quick donning, Cost effective protection, Size: 3xl
Catalog #: 76486-274
Supplier:  PIP
Lab Coat with No Pocket, SBP, Blue, polypropylene disposable clothing provides affordable, lightweight protection against non-hazardous dry particulate like dirt, Professional collar, Full Length coverage to top of thigh, Snap button for front for quick donning, Cost effective protection, Size: 3xl
Catalog #: 76486-274
Supplier:  PIP
Customers Who Bought This Also Bought