ContactUsLinkComponent
Coverall with Elastic Wrist & Ankle with Attached Hood & Boot, PE Laminate in PP provides lightweight, breathable, protection against non hazardous light spray, splashes in manufacturing, Full length zipper front for easy donning, Open wrist/ankle, Latex Free, White, One time use, Size: 2xl
Catalog #: 76486-194
Supplier:  PIP
Coverall with Elastic Wrist & Ankle with Attached Hood & Boot, PE Laminate in PP provides lightweight, breathable, protection against non hazardous light spray, splashes in manufacturing, Full length zipper front for easy donning, Open wrist/ankle, Latex Free, White, One time use, Size: 2xl
Catalog #: 76486-194
Supplier:  PIP
Customers Who Bought This Also Bought