ContactUsLinkComponent
Coverall Hood with Elastic Wrist & Ankle, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light spray, splashes in manufacturing, mold remediation, Full length zipper front for easy donning, Open wrist/ankle, Latex Free, White, One time use, Size: 3XL
Catalog #: 76486-182
Supplier:  PIP
Coverall Hood with Elastic Wrist & Ankle, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light spray, splashes in manufacturing, mold remediation, Full length zipper front for easy donning, Open wrist/ankle, Latex Free, White, One time use, Size: 3XL
Catalog #: 76486-182
Supplier:  PIP
Customers Who Bought This Also Bought