ContactUsLinkComponent
Coverall, with Elastic Wrist & Ankle, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light sprays, splashes in manufacturing, mold remediation, Full length zipper front for easy donning, Open wrist/ankles, Latex Free, White, One time use, Size: 4xl
Catalog #: 76486-170
Supplier:  PIP
Coverall, with Elastic Wrist & Ankle, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light sprays, splashes in manufacturing, mold remediation, Full length zipper front for easy donning, Open wrist/ankles, Latex Free, White, One time use, Size: 4xl
Catalog #: 76486-170
Supplier:  PIP
Customers Who Bought This Also Bought