ContactUsLinkComponent
Coverall, Basic, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light sprays, splashes in manufacturing, mold remediation, cleaning, Full length zipper front for easy donning, Open wrist/ankles, Latex Free, White in color, One time use, Size: 5xl
Catalog #: 76486-160
Supplier:  PIP
Coverall, Basic, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light sprays, splashes in manufacturing, mold remediation, cleaning, Full length zipper front for easy donning, Open wrist/ankles, Latex Free, White in color, One time use, Size: 5xl
Catalog #: 76486-160
Supplier:  PIP
Customers Who Bought This Also Bought