Coverall, Basic, PE Laminate in PP provides lightweight, breathable, comfortable protection against non hazardous light sprays, splashes in manufacturing, mold remediation, cleaning, Full length zipper front for easy donning, Open wrist/ankles, Latex Free, White in color, One time use, Size: 4xl
Catalog #: 76486-158
Supplier: PIP
Customers Who Bought This Also Bought