ContactUsLinkComponent
Coverall, Posi-Wear Flame Resistant with Hood, Boot, Elastic Wrists and Ankles, Meets NFPA 701 specifications for Self- Extinguishing, Anti-stat and ANSI Sized, Latex Free, Blue, Base Material: Spunlace Self- Extinguishing, Designed for use in applications where sparks are present, Size: 2xl
Catalog #: 76486-136
Supplier:  PIP
Coverall, Posi-Wear Flame Resistant with Hood, Boot, Elastic Wrists and Ankles, Meets NFPA 701 specifications for Self- Extinguishing, Anti-stat and ANSI Sized, Latex Free, Blue, Base Material: Spunlace Self- Extinguishing, Designed for use in applications where sparks are present, Size: 2xl
Catalog #: 76486-136
Supplier:  PIP
Customers Who Bought This Also Bought