ContactUsLinkComponent
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, blue, pack of 5
Microcentrifuge Tube Racks
Catalog #: 470334-390
Supplier:  MTC BIO, INC.
Microcentrifuge Tube Racks
Catalog #: 470334-390
Supplier:  MTC BIO, INC.
Specifications
  • Color:
    Blue
  • Description:
    Microcentrifuge tube racks
  • No. of Holes:
    80
  • Cat. no.:
    470334-390
  • For Tubes:
    1.5/2.0 ml
Specifications
About this item
Compact microcentrifuge tube racks to keep you and your samples organized.
  • Autoclavable
  • Accomodates conical and self-standing tubes
  • Raised, molded numbers
These compact, convenient 80 well racks are designed to keep your tubes neatly and efficiently organized. They are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification.
The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes. They are made of autoclavable polypropylene and are supplied in a rainbow pack of five racks (B, O, R, G, Y) or a single color of your choice.
Customers Who Bought This Also Bought