FreezeBox Extended Height CF, Blue, Package includes 1 each of FreezeBox Extended Height, CF Freeze Core -18C to -4C, and CM-03 Tube Module for 30 x 5mL tubes 60mm, 152 x 170 x 170mm, Uniform, low temperature protection for consistent sample results, typically +/-0.1 C or taller
Catalog #: 76480-468
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Customers Who Bought This Also Bought